Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142952.3 | NP_651209.1 | 1 - 2 | 255 | 72 | |
GenBank mRNA | AY061424.1 | - | 1 - 2 | 486 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |