Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142963.3 | NP_651220.3 | 2 - 3 | 944 | 68 | |
GenBank mRNA | AF342988.1 | - | 2 - 3 | 907 | 68 | |
BT015965.1 | - | 2 - 3 | 945 | 68 | ||
BT044162.1 | - | 2 - 3 | 899 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |