Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300577.1 | NP_001287506.1 | 3 - 4 | 1685 | 74 | |
NM_143017.3 | NP_651274.1 | 3 - 4 | 1667 | 74 | ||
NM_170157.2 | NP_733020.1 | 3 - 4 | 1985 | 74 | ||
GenBank mRNA | AY069079.1 | - | 3 - 4 | 1620 | 74 | |
BT004893.1 | - | 3 - 4 | 1980 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |