Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170242.2 | NP_001163713.1 | 2 - 3 | 207 | 92 | |
NM_001276003.1 | NP_001262932.1 | 2 - 3 | 207 | 92 | ||
NM_143040.5 | NP_651297.2 | 2 - 3 | 207 | 92 | ||
NM_206568.4 | NP_996291.2 | 2 - 3 | 207 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |