Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170242.2 | NP_001163713.1 | 6 - 7 | 2127 | 55 | |
NM_143040.5 | NP_651297.2 | 6 - 7 | 2127 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |