Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170250.1 | NP_001163721.1 | 4 - 5 | 2955 | 88 | |
NM_143084.2 | NP_651341.2 | 5 - 6 | 2943 | 88 | ||
GenBank mRNA | BT030842.1 | - | 4 - 5 | 2893 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |