Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bili | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170251.2 | NP_001163722.1 | 3 - 4 | 577 | 90 | |
NM_001170252.2 | NP_001163723.1 | 3 - 4 | 566 | 90 | ||
NM_001170253.2 | NP_001163724.1 | 3 - 4 | 661 | 90 | ||
NM_001170254.2 | NP_001163725.1 | 3 - 4 | 597 | 90 | ||
NM_001170255.1 | NP_001163726.1 | 3 - 4 | 717 | 90 | ||
NM_001300592.1 | NP_001287521.1 | 3 - 4 | 970 | 90 | ||
NM_143085.3 | NP_651342.3 | 3 - 4 | 674 | 90 | ||
GenBank mRNA | AY094737.1 | - | 3 - 4 | 636 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |