Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKov2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143130.4 | NP_651387.1 | 1 - 2 | 228 | 96 | |
GenBank mRNA | AY058384.1 | - | 1 - 2 | 207 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |