Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143197.2 | NP_651454.1 | 4 - 5 | 1326 | 62 | |
NM_206576.2 | NP_996299.1 | 4 - 5 | 1329 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |