Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276084.1 | NP_001263013.1 | 3 - 4 | 1023 | 65 | |
NM_143314.4 | NP_651571.3 | 3 - 4 | 1023 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |