Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202374.2 | NP_001189303.1 | 9 - 10 | 4787 | 78 | |
NM_001276085.2 | NP_001263014.1 | 9 - 10 | 4424 | 78 | ||
NM_143316.4 | NP_651573.4 | 9 - 10 | 4187 | 78 | ||
GenBank mRNA | BT132902.1 | - | 9 - 10 | 4187 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |