Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276088.1 | NP_001263017.1 | 1 - 2 | 798 | 78 | |
NM_001276089.1 | NP_001263018.1 | 1 - 2 | 616 | 78 | ||
NM_001276090.2 | NP_001263019.2 | 1 - 2 | 616 | 78 | ||
NM_001300634.1 | NP_001287563.1 | 1 - 2 | 616 | 78 | ||
NM_001300635.1 | NP_001287564.1 | 1 - 2 | 616 | 78 | ||
NM_001300636.1 | NP_001287565.1 | 1 - 2 | 616 | 78 | ||
NM_001300639.1 | NP_001287568.1 | 1 - 2 | 616 | 78 | ||
NM_001316620.1 | NP_001303549.1 | 1 - 2 | 616 | 78 | ||
NM_001382126.1 | NP_001369032.1 | 1 - 2 | 616 | 78 | ||
NM_143318.3 | NP_651575.1 | 1 - 2 | 616 | 78 | ||
GenBank mRNA | AY032977.1 | - | 1 - 2 | 593 | 78 | |
AY102697.1 | - | 1 - 2 | 612 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |