Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143343.3 | NP_651600.1 | 3 - 4 | 1039 | 87 | |
GenBank mRNA | AY060268.1 | - | 3 - 4 | 1019 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |