Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276116.1 | NP_001263045.1 | 6 - 7 | 1536 | 91 | |
NM_143380.4 | NP_651637.1 | 6 - 7 | 1596 | 91 | ||
NM_170378.2 | NP_733257.1 | 6 - 7 | 1484 | 91 | ||
NM_176578.2 | NP_788755.1 | 5 - 6 | 1302 | 91 | ||
GenBank mRNA | AY070576.1 | - | 6 - 7 | 1607 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |