Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276153.1 | NP_001263082.1 | 3 - 4 | 1577 | 99 | |
NM_143465.2 | NP_651722.1 | 3 - 4 | 1577 | 99 | ||
GenBank mRNA | BT032863.1 | - | 3 - 4 | 1561 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |