Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143502.4 | NP_651759.4 | 4 - 5 | 1286 | 87 | |
NM_170455.3 | NP_733334.1 | 6 - 7 | 1372 | 87 | ||
GenBank mRNA | AY212802.1 | - | 6 - 7 | 1348 | 87 | |
BT003184.1 | - | 6 - 7 | 1348 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |