Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276180.1 | NP_001263109.1 | 2 - 3 | 176 | 73 | |
NM_143540.2 | NP_651797.1 | 2 - 3 | 479 | 73 | ||
NM_170488.2 | NP_733367.1 | 2 - 3 | 212 | 73 | ||
NM_170489.2 | NP_733368.1 | 2 - 3 | 584 | 73 | ||
NM_170490.2 | NP_733369.1 | 2 - 3 | 580 | 73 | ||
NM_170491.2 | NP_733370.1 | 3 - 4 | 715 | 73 | ||
GenBank mRNA | BT150409.1 | - | 2 - 3 | 514 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |