Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260210.2 | NP_001247139.1 | 1 - 2 | 261 | 65 | |
NM_144354.3 | NP_652611.1 | 1 - 2 | 261 | 65 | ||
GenBank mRNA | AF232773.1 | - | 1 - 2 | 245 | 65 | |
AY051919.1 | - | 1 - 2 | 245 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |