Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260210.2 | NP_001247139.1 | 3 - 4 | 754 | 128 | |
NM_144354.3 | NP_652611.1 | 3 - 4 | 754 | 128 | ||
GenBank mRNA | AF232773.1 | - | 3 - 4 | 738 | 128 | |
AY051919.1 | - | 3 - 4 | 738 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |