Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG46339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104469.3 | NP_001097939.1 | 10 - 11 | 3794 | 70 | |
NM_001260394.2 | NP_001247323.1 | 10 - 11 | 3497 | 70 | ||
NM_001276072.1 | NP_001263001.1 | 11 - 12 | 3830 | 70 | ||
NM_001363935.1 | NP_001350863.1 | 8 - 9 | 3365 | 70 | ||
NM_001363936.1 | NP_001350864.1 | 9 - 10 | 3698 | 70 | ||
NM_144487.3 | NP_652744.2 | 9 - 10 | 3461 | 70 | ||
GenBank mRNA | AF480087.1 | - | 9 - 10 | 3438 | 70 | |
AY122083.1 | - | 9 - 10 | 3501 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |