Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080126.4 | NP_524865.2 | 2 - 3 | 360 | 58 | |
NM_164321.5 | NP_730790.2 | 3 - 4 | 554 | 58 | ||
NM_176394.3 | NP_788571.1 | 2 - 3 | 331 | 58 | ||
NM_206427.4 | NP_996149.3 | 3 - 4 | 421 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |