Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275345.1 | NP_001262274.1 | 16 | 6024 | 95 | |
NM_169026.3 | NP_730892.2 | 16 | 6021 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |