Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmdar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169059.2 | NP_730940.1 | 3 - 4 | 544 | 76 | |
GenBank mRNA | AY070577.1 | - | 3 - 4 | 540 | 76 | |
X71790.1 | - | 452 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |