Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4KIIalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382052.1 | NP_001368956.1 | 6 - 7 | 2005 | 75 | |
NM_001382053.1 | NP_001368992.1 | 5 - 6 | 1331 | 75 | ||
NM_169063.4 | NP_730944.2 | 7 - 8 | 2409 | 75 | ||
NM_169064.2 | NP_730945.1 | 5 - 6 | 1852 | 75 | ||
NM_206432.3 | NP_996154.1 | 7 - 8 | 2382 | 75 | ||
GenBank mRNA | AY102687.1 | - | 7 - 8 | 2355 | 75 | |
BT001304.1 | - | 5 - 6 | 1852 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |