Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169068.3 | NP_730950.2 | 2 - 3 | 424 | 60 | |
NM_169069.3 | NP_730951.2 | 3 - 4 | 258 | 60 | ||
GenBank mRNA | AY094697.1 | - | 3 - 4 | 220 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |