Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170040.2 | NP_001163511.1 | 5 - 6 | 2130 | 90 | |
NM_169094.2 | NP_730988.1 | 2 - 3 | 1370 | 90 | ||
NM_169095.3 | NP_730989.2 | 3 - 4 | 1277 | 90 | ||
GenBank mRNA | BT133115.1 | - | 5 - 6 | 1853 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |