Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfzf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014609.3 | NP_001014609.2 | 1 - 2 | 49 | 63 | |
GenBank mRNA | BT012465.1 | - | 1 - 2 | 46 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |