Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jupiter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169405.4 | NP_731596.1 | 3 - 4 | 596 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |