Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jupiter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169406.4 | NP_731597.1 | 3 - 4 | 365 | 74 | |
NM_169408.4 | NP_731599.1 | 4 - 5 | 473 | 74 | ||
GenBank mRNA | BT001513.1 | - | 3 - 4 | 342 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |