Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300370.1 | NP_001287299.1 | 5 - 6 | 2069 | 80 | |
NM_169453.1 | NP_731672.1 | 5 - 6 | 2085 | 80 | ||
GenBank mRNA | BT044240.1 | - | 5 - 6 | 2085 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |