Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-Vb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275582.1 | NP_001262511.1 | 3 - 4 | 569 | 97 | |
NM_169481.1 | NP_731748.1 | 3 - 4 | 764 | 97 | ||
GenBank mRNA | AY070836.1 | - | 764 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |