Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Act87E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300385.1 | NP_001287314.1 | 2 | 1477 | 89 | |
NM_057743.5 | NP_477091.1 | 2 | 1477 | 89 | ||
NM_169525.2 | NP_731812.1 | 1 | 1553 | 89 | ||
GenBank mRNA | AY089587.1 | - | 2 | 1475 | 89 | |
BT016152.1 | - | 1 | 1553 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |