Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stumps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104322.2 | NP_001097792.1 | 4 - 5 | 892 | 61 | |
GenBank mRNA | BT024930.1 | - | 4 - 5 | 890 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |