Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hel89B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300422.1 | NP_001287351.1 | 4 - 5 | 3202 | 60 | |
NM_169693.2 | NP_732097.2 | 4 - 5 | 3629 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |