Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mhcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043250.2 | NP_001036715.1 | 6 - 7 | 2194 | 77 | |
NM_001043251.2 | NP_001036716.1 | 3 - 4 | 1921 | 77 | ||
NM_169700.3 | NP_732108.2 | 6 - 7 | 2194 | 77 | ||
NM_169701.3 | NP_732109.2 | 6 - 7 | 2194 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |