Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104355.2 | NP_001097825.1 | 32 - 33 | 6626 | 82 | |
NM_169761.2 | NP_732242.2 | 32 - 33 | 6149 | 82 | ||
NM_206504.2 | NP_996226.2 | 32 - 33 | 6149 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |