Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043268.2 | NP_001036733.1 | 5 - 6 | 1174 | 79 | |
GenBank mRNA | BT056325.1 | - | 5 - 6 | 1172 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |