Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oamb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275845.2 | NP_001262774.1 | 6 - 7 | 2008 | 93 | |
NM_001316500.1 | NP_001303429.1 | 6 - 7 | 2008 | 93 | ||
NM_169913.2 | NP_732541.1 | 6 - 7 | 2008 | 93 | ||
NM_169914.2 | NP_732542.1 | 5 - 6 | 1582 | 93 | ||
GenBank mRNA | AJ007617.1 | - | 4 - 5 | 1471 | 93 | |
BT010103.1 | - | 6 - 7 | 1972 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |