Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043281.2 | NP_001036746.1 | 10 - 11 | 5328 | 56 | |
NM_001275918.1 | NP_001262847.1 | 10 - 11 | 5334 | 56 | ||
NM_170037.2 | NP_732790.3 | 10 - 11 | 5361 | 56 | ||
NM_170038.2 | NP_732791.2 | 9 - 10 | 5184 | 56 | ||
GenBank mRNA | AB190214.1 | - | 10 - 11 | 4734 | 56 | |
BT099586.1 | - | 10 - 11 | 5315 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |