Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043281.2 | NP_001036746.1 | 8 - 9 | 4987 | 71 | |
NM_001275918.1 | NP_001262847.1 | 8 - 9 | 4993 | 71 | ||
NM_170037.2 | NP_732790.3 | 8 - 9 | 5020 | 71 | ||
NM_170038.2 | NP_732791.2 | 7 - 8 | 4843 | 71 | ||
GenBank mRNA | AB190214.1 | - | 8 - 9 | 4393 | 71 | |
BT099586.1 | - | 8 - 9 | 4974 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |