Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CenB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170049.4 | NP_732826.2 | 2 - 3 | 339 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |