Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104422.3 | NP_001097892.1 | 5 - 6 | 1645 | 81 | |
NM_001104423.2 | NP_001097893.1 | 5 - 6 | 1574 | 81 | ||
NM_001260342.2 | NP_001247271.1 | 5 - 6 | 1574 | 81 | ||
NM_001275959.1 | NP_001262888.1 | 5 - 6 | 1574 | 81 | ||
NM_001275960.1 | NP_001262889.1 | 5 - 6 | 1645 | 81 | ||
NM_170094.4 | NP_732911.2 | 4 - 5 | 1382 | 81 | ||
NM_176549.3 | NP_788726.2 | 4 - 5 | 1382 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |