Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KrT95D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170238.2 | NP_001163709.2 | 1 - 2 | 1332 | 82 | |
NM_079749.6 | NP_524473.2 | 1 - 2 | 1332 | 82 | ||
NM_170108.4 | NP_732933.1 | 2 - 3 | 652 | 82 | ||
NM_170110.4 | NP_732935.2 | 2 - 3 | 652 | 82 | ||
GenBank mRNA | AY119444.1 | - | 2 - 3 | 635 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |