Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KrT95D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170238.2 | NP_001163709.2 | 4 - 5 | 2226 | 70 | |
NM_079749.6 | NP_524473.2 | 4 - 5 | 2226 | 70 | ||
NM_170108.4 | NP_732933.1 | 5 - 6 | 1546 | 70 | ||
NM_170109.2 | NP_732934.2 | 2 - 3 | 562 | 70 | ||
NM_170110.4 | NP_732935.2 | 5 - 6 | 1546 | 70 | ||
GenBank mRNA | AY061587.1 | - | 3 - 4 | 450 | 70 | |
AY119444.1 | - | 5 - 6 | 1529 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |