Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170155.2 | NP_733018.1 | 1 - 2 | 945 | 82 | |
GenBank mRNA | BT001869.1 | - | 1 - 2 | 960 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |