Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RabX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170170.3 | NP_733043.1 | 1 - 2 | 577 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |