Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170236.2 | NP_733115.1 | 6 - 7 | 1432 | 83 | |
GenBank mRNA | BT150256.1 | - | 6 - 7 | 1432 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |