Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LpR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104456.2 | NP_001097926.1 | 10 - 11 | 3281 | 74 | |
NM_001104457.2 | NP_001097927.1 | 6 - 7 | 1334 | 74 | ||
NM_001104458.2 | NP_001097928.1 | 11 - 12 | 3386 | 74 | ||
NM_001104459.2 | NP_001097929.1 | 10 - 11 | 2845 | 74 | ||
NM_001104460.2 | NP_001097930.1 | 11 - 12 | 3383 | 74 | ||
NM_001276041.1 | NP_001262970.1 | 10 - 11 | 2834 | 74 | ||
NM_170239.4 | NP_733118.3 | 10 - 11 | 3278 | 74 | ||
NM_176565.2 | NP_788742.1 | 9 - 10 | 2673 | 74 | ||
GenBank mRNA | BT010074.1 | - | 11 - 12 | 3364 | 74 | |
BT010094.1 | - | 10 - 11 | 2756 | 74 | ||
BT011437.1 | - | 6 - 7 | 1335 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster Inventoried |