Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LpR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104461.3 | NP_001097931.3 | 2 - 3 | 325 | 67 | |
NM_001104462.1 | NP_001097932.1 | 2 - 3 | 325 | 67 | ||
NM_170240.3 | NP_733119.2 | 2 - 3 | 325 | 67 | ||
GenBank mRNA | BT099946.1 | - | 2 - 3 | 325 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |