Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202364.1 | NP_001189293.1 | 11 - 12 | 2378 | 70 | |
NM_001202365.2 | NP_001189294.1 | 12 - 13 | 2399 | 70 | ||
NM_001260380.2 | NP_001247309.1 | 11 - 12 | 2342 | 70 | ||
NM_001276052.1 | NP_001262981.1 | 10 - 11 | 2309 | 70 | ||
NM_001300610.1 | NP_001287539.1 | 11 - 12 | 2366 | 70 | ||
NM_079790.5 | NP_524514.2 | 11 - 12 | 2311 | 70 | ||
NM_170254.3 | NP_733133.1 | 11 - 12 | 2391 | 70 | ||
NM_170255.4 | NP_733134.1 | 11 - 12 | 2372 | 70 | ||
NM_170256.2 | NP_733135.1 | 11 - 12 | 2290 | 70 | ||
NM_206575.3 | NP_996298.1 | 12 - 13 | 2405 | 70 | ||
GenBank mRNA | AF145695.1 | - | 11 - 12 | 2357 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |